accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOCH

CAS RN 23418-85-1

IUPAC Name: but-3-yn-1-yl 4-methylbenzene-1-sulfonate
Synonyms: but-3-yn-1-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 224.27
Molecular Formula: C11H12O3S
InChi Key: STOASOOVVADOKH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCC#C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)