accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2346-81-8

IUPAC Name: 3-chlorohexane
Synonyms: hexane, 3-chloro-
Molecular Weight (g/mol): 120.62
Molecular Formula: C6H13Cl
InChi Key: BXSMMAVTEURRGG-UHFFFAOYNA-N
SMILES: CCCC(Cl)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results