accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOO

CAS RN 23489-36-3

IUPAC Name: 1-(1-benzofuran-2-yl)-2-bromoethan-1-one
Synonyms: 1-(1-benzofuran-2-yl)-2-bromoethanone
Molecular Weight (g/mol): 239.07
Molecular Formula: C10H7BrO2
InChi Key: NVRNCBWTEDOAQA-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results