accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 235-92-7

IUPAC Name: 1H-cyclopenta[l]phenanthrene
Synonyms: 1H-cyclopenta[l]phenanthrene
Molecular Weight (g/mol): 216.28
Molecular Formula: C17H12
InChi Key: YQDSWSPIWVQKBC-UHFFFAOYSA-N
SMILES: C1C=CC2=C1C1=CC=CC=C1C1=CC=CC=C21
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results