Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Cyclopenta[l]phenanthrene, TCI America™
SDP

Catalog No. C16891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16891G Supplier TCI America Supplier No. C16891G

missing translation for 'validMainframeMsg'

Specifications

CAS 235-92-7
Melting Point 151°C
Molecular Formula C17H12
Quantity 1 g
MDL Number MFCD03844783
InChI Key YQDSWSPIWVQKBC-UHFFFAOYSA-N
SMILES C1C=CC2=C1C3=CC=CC=C3C4=CC=CC=C24
IUPAC Name 1H-cyclopenta[l]phenanthrene
Molecular Weight (g/mol) 216.283
PubChem CID 11806107
Formula Weight 216.28
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H-Cyclopenta[l]phenanthrene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.