accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2350-01-8

IUPAC Name: N1,N1-diphenylbenzene-1,4-diamine
Synonyms: N1,N1-diphenylbenzene-1,4-diamine
Molecular Weight (g/mol): 260.34
Molecular Formula: C18H16N2
InChi Key: UXKQNCDDHDBAPD-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results