Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminotriphenylamine 95.0+%, TCI America™
SDP

Catalog No. A2624200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2624200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2624200MG Supplier TCI America Supplier No. A2624200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2350-01-8
Molecular Formula C18H16N2
Molecular Weight (g/mol) 260.34
MDL Number MFCD00145130
InChI Key UXKQNCDDHDBAPD-UHFFFAOYSA-N
Synonym N,N-Diphenyl-1,4-phenylenediamine, 4-Amino-N,N-diphenylaniline
PubChem CID 75371
IUPAC Name 4-N,4-N-diphenylbenzene-1,4-diamine
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)N

Specifications

Melting Point 146°C
Boiling Point 270°C
Quantity 200 mg
Formula Weight 260.34
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminotriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.