CAS RN 2351-33-9
IUPAC Name:
1,1-dichlorosiletane
Synonyms:
1,1-dichlorosiletane
Molecular Weight (g/mol):
141.07
Molecular Formula:
C3H6Cl2Si
InChi Key:
PASYEMKYRSIVTP-UHFFFAOYSA-N
SMILES:
Cl[Si]1(Cl)CCC1
1
–
1
of
1
results
1
Sigma Aldrich Lead(II) Tetrafluoroborate Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.