Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lead(II) Tetrafluoroborate Solution

Catalog No. 401579250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-157-9250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 40-157-9250ML Supplier Sigma Aldrich Supplier No. 401579250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for mediation of calixarene formation via hydrogen-bonded dimersAnalyzed in terms of electochemical characterization of the effects of impurities and organic additives in lead electrowinning from fluoroborate electrolyteUsed in the development and optimization of a bipolar lead-boron tetrafluoride accumulator

Specifications

CAS 2351-33-9
Density 1.19 g/mL at 25°C
Boiling Point 113°C to 115°C
Molecular Formula C3H6Cl2Si
Linear Formula C3H6Cl2Si
Quantity 250 mL
MDL Number MFCD00053537
Synonym Lead boron fluoride; Lead(II) fluoroborate
Molecular Weight (g/mol) 141.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.