accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHSNHNHSNHOOCH3

CAS RN 23564-05-8

IUPAC Name: methyl N-{[2-({[(methoxycarbonyl)amino]methanethioyl}amino)phenyl]carbamothioyl}carbamate
Synonyms: zyban fungo thiophanate-methyl
Molecular Weight (g/mol): 342.39
Molecular Formula: C12H14N4O4S2
InChi Key: QGHREAKMXXNCOA-UHFFFAOYSA-N
SMILES: COC(=O)NC(=S)NC1=CC=CC=C1NC(=S)NC(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results