CAS RN 2357-47-3
CAS RN 2357-47-3
IUPAC Name:
4-fluoro-3-(trifluoromethyl)aniline
Synonyms:
4-fluoro-3-(trifluoromethyl)aniline
Molecular Weight (g/mol):
179.12
Molecular Formula:
C7H5F4N
InChi Key:
PGFQDLOMDIBAPY-UHFFFAOYSA-N
SMILES:
NC1=CC=C(F)C(=C1)C(F)(F)F
1
–
1
of
1
results
1
5-Amino-2-fluorobenzotrifluoride 98.0+%, TCI America™
CAS: 2357-47-3 Molecular Formula: C7H5F4N Molecular Weight (g/mol): 179.118 MDL Number: MFCD00007834 InChI Key: PGFQDLOMDIBAPY-UHFFFAOYSA-N Synonym: 4-fluoro-3-trifluoromethyl aniline,5-amino-2-fluorobenzotrifluoride,alpha,alpha,alpha,4-tetrafluoro-m-toluidine,3-trifluoromethyl-4-fluoroaniline,4-fluoro-3-trifluoromethyl benzenamine,benzenamine, 4-fluoro-3-trifluoromethyl,unii-v5584x2ctl,2-fluoro-5-aminobenzotrifluoride,5-amino-2-fluoro trifluoromethyl benzene,4-fluoro-3-trifluoromethyl phenylamine PubChem CID: 75377 IUPAC Name: 4-fluoro-3-(trifluoromethyl)aniline SMILES: C1=CC(=C(C=C1N)C(F)(F)F)F