Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-fluorobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. A11645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11645G 5 g
A116425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11645G Supplier TCI America Supplier No. A11645G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2357-47-3
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.118
MDL Number MFCD00007834
InChI Key PGFQDLOMDIBAPY-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl aniline, 5-amino-2-fluorobenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-m-toluidine, 3-trifluoromethyl-4-fluoroaniline, 4-fluoro-3-trifluoromethyl benzenamine, benzenamine, 4-fluoro-3-trifluoromethyl, unii-v5584x2ctl, 2-fluoro-5-aminobenzotrifluoride, 5-amino-2-fluoro trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl phenylamine
PubChem CID 75377
IUPAC Name 4-fluoro-3-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1N)C(F)(F)F)F

Specifications

Color Yellow
Boiling Point 208°C
Quantity 5 g
Formula Weight 179.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Amino-2-fluorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.