accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2358-29-4

IUPAC Name: 2,3,6-trifluorobenzoate
Synonyms: 2,3,6-trifluorobenzoate
Molecular Weight (g/mol): 175.09
Molecular Formula: C7H2F3O2
InChi Key: MGUPHQGQNHDGNK-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C(F)C=CC(F)=C1F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)