Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6-Trifluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. T14565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14565G 5 g
T145625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14565G Supplier TCI America Supplier No. T14565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2358-29-4
Molecular Formula C7H2F3O2
Molecular Weight (g/mol) 175.09
MDL Number MFCD00002409
InChI Key MGUPHQGQNHDGNK-UHFFFAOYSA-M
Synonym 2,5,6-trifluorobenzoic acid, 2,3,6-trifluorobenzoicacid, 2,3,6-trifluoro-benzoic acid, benzoic acid, 2,3,6-trifluoro, pubchem1344, intermediates-zcf02628, ksc494k2n, 2,3,6-trifluoro benzoic acid, rarechem al bo 0267, 2,3,6-trifluorobenzoic acid
PubChem CID 520062
IUPAC Name 2,3,6-trifluorobenzoate
SMILES [O-]C(=O)C1=C(F)C=CC(F)=C1F

Specifications

Melting Point 129°C
Color White
Quantity 5 g
Formula Weight 176.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6-Trifluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.