accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2358-84-1

H3CCH2OOOOOCH3CH2

CAS RN 2358-84-1

IUPAC Name: 2-{2-[(2-methylprop-2-enoyl)oxy]ethoxy}ethyl 2-methylprop-2-enoate
Synonyms: diethylene glycol dimethacrylate
Molecular Weight (g/mol): 242.27
Molecular Formula: C12H18O5
InChi Key: XFCMNSHQOZQILR-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCOCCOC(=O)C(C)=C
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)