Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di(ethylene glycol) dimethacrylate

Catalog No. 409006250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-900-6250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-900-6250ML Supplier Sigma Aldrich Supplier No. 409006250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Di(ethylene glycol) dimethacrylate

Specifications

CAS 2358-84-1
Density 1.082 g/mL (at 25°C)
Boiling Point 134°C (2 mmHg)
Molecular Formula C12H18O5
Refractive Index n20/D 1.458 (literature)
Linear Formula [H2C=C(CH3)CO2CH2CH2]2 O
Quantity 250 mL
MDL Number MFCD00014931
Synonym 2,2 -Oxybisethanol dimethacrylate; 2,2 -Oxydiethyl dimethacrylate; Diethylene glycol, dimethacrylate
Molecular Weight (g/mol) 242.27
Percent Purity 95%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.