accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23583-48-4

IUPAC Name: (4aR,6R,7R,7aS)-6-(6-amino-8-bromo-9H-purin-9-yl)-2,7-dihydroxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
Synonyms: 8-Br-cAMP 8-bromo-cyclic amp 8-bromo-cyclic 3',5'-amp 8-bromo-3',5'-cyclic amp bcamp
Molecular Weight (g/mol): 408.11
Molecular Formula: C10H11BrN5O6P
InChi Key: DVKQVRZMKBDMDH-UUOKFMHZSA-N
SMILES: NC1=C2N=C(Br)N([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)