accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2359-15-1

IUPAC Name: 2-methyl-N-[(2-methylprop-2-enamido)methyl]prop-2-enamide
Synonyms: 2-methyl-N-[(2-methylprop-2-enamido)methyl]prop-2-enamide
Molecular Weight (g/mol): 182.22
Molecular Formula: C9H14N2O2
InChi Key: TURITJIWSQEMDB-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)NCNC(=O)C(C)=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results