Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Methylenebismethacrylamide 96.0+%, TCI America™
SDP

Catalog No. M093025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M093025G 25 g
M0930500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M093025G Supplier TCI America Supplier No. M093025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2359-15-1
Molecular Formula C9H14N2O2
Molecular Weight (g/mol) 182.223
MDL Number MFCD00060096
InChI Key TURITJIWSQEMDB-UHFFFAOYSA-N
PubChem CID 75381
IUPAC Name 2-methyl-N-[(2-methylprop-2-enoylamino)methyl]prop-2-enamide
SMILES CC(=C)C(=O)NCNC(=O)C(=C)C

Specifications

Color White
Quantity 25 g
Formula Weight 182.22
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Methylenebismethacrylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.