accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOOHCH3

CAS RN 2362-14-3

IUPAC Name: 4-[1-(4-hydroxy-3-methylphenyl)cyclohexyl]-2-methylphenol
Synonyms: 4-[1-(4-hydroxy-3-methylphenyl)cyclohexyl]-2-methylphenol
Molecular Weight (g/mol): 296.41
Molecular Formula: C20H24O2
InChi Key: SVOBELCYOCEECO-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1O)C1(CCCCC1)C1=CC=C(O)C(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results