Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Bis(4-hydroxy-3-methylphenyl)cyclohexane 98.0+%, TCI America™
SDP

Catalog No. B275325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B275325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B275325G Supplier TCI America Supplier No. B275325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2362-14-3
Molecular Formula C20H24O2
Molecular Weight (g/mol) 296.41
InChI Key SVOBELCYOCEECO-UHFFFAOYSA-N
Synonym 4,4′C-Cyclohexylidenebis(o-cresol), 4,4′C-Cyclohexylidenebis(2-methylphenol)
PubChem CID 75386
IUPAC Name 4-[1-(4-hydroxy-3-methylphenyl)cyclohexyl]-2-methylphenol
SMILES CC1=C(C=CC(=C1)C2(CCCCC2)C3=CC(=C(C=C3)O)C)O

Specifications

Melting Point 191°C
Color White
Quantity 25 g
Formula Weight 296.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Bis(4-hydroxy-3-methylphenyl)cyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.