accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2364-59-2

IUPAC Name: ethyl 4-phenoxybutanoate
Synonyms: ethyl 4-phenoxybutanoate
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: XSVIVXBQBFMYCS-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCOC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results