Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Phenoxybutyrate 96.0+%, TCI America™
SDP

Catalog No. P128625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P128625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P128625G Supplier TCI America Supplier No. P128625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2364-59-2
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD01321182
InChI Key XSVIVXBQBFMYCS-UHFFFAOYSA-N
Synonym 4-Phenoxybutyric Acid Ethyl Ester
PubChem CID 222539
IUPAC Name ethyl 4-phenoxybutanoate
SMILES CCOC(=O)CCCOC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 173°C
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Phenoxybutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.