CAS RN 236403-25-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ MMP-8 Inhibitor I
The MMP-8 Inhibitor I, also referenced under CAS 236403-25-1, controls the biological activity of MMP-8. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.