Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MMP-8 Inhibitor I

Catalog No. 4442371MG Shop All MilliporeSigma Products
Change view
Click to view available options
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
44-423-71MG 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-423-71MG Supplier MilliporeSigma™ Supplier No. 4442371MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

The MMP-8 Inhibitor I, also referenced under CAS 236403-25-1, controls the biological activity of MMP-8. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins MMP-8
CAS 236403-25-1
Molecular Formula C17H18N2O5S
Formula Weight 362.4g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.