accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2365-48-2

IUPAC Name: methyl 2-sulfanylacetate
Synonyms: methyl thioglycolate
Molecular Weight (g/mol): 106.14
Molecular Formula: C3H6O2S
InChi Key: MKIJJIMOAABWGF-UHFFFAOYSA-N
SMILES: COC(=O)CS
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (5)
  • (2)
Quantity 
  • (4)
  • (1)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (5)