Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl thioglycolate

Catalog No. 1089955G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-899-55G 5 g
10-899-5500G 500 g
10-899-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-899-55G Supplier Sigma Aldrich Supplier No. 1089955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl thioglycolate reacts with nonprotein component of the antitumor antibiotic neocarzinostatin to form 1:1 adduct. it reacts with isothiocyanate to form rhodanine.methyl thioglycolate was used in the preparation of:
  • 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene
  • Methyl thioglycolate and aminoethanethiol conjugated gold nanorods.thiol organocatalyst used in conjunction with visible light photocatalysis to affect the direct arylation of benzylic ether c-h bonds a general strategy for organocatalytic activation of c-h bonds via photoredox catalysis: direct arylation of benzylic ethers

Specifications

CAS 2365-48-2
Density 1.187 g/mL (at 25°C (literature))
Boiling Point 42°C to 43°C (10 mmHg)
Molecular Formula C3H6O2S
Refractive Index n20/D 1.466 (literature)
Linear Formula HSCH2CO2CH3
MDL Number MFCD00004873
Quantity 5 g
Synonym Methyl mercaptoacetate
Molecular Weight (g/mol) 106.14
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.