accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFFF

CAS RN 2366-70-3

IUPAC Name: 4,4,4-trifluorobutan-2-one
Synonyms: 4,4,4-trifluorobutan-2-one
Molecular Weight (g/mol): 126.08
Molecular Formula: C4H5F3O
InChi Key: BTXXTMOWISPQSJ-UHFFFAOYSA-N
SMILES: CC(=O)CC(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)