accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(R)O(R)(S)HONONHOOCH3

CAS RN 23669-79-6

IUPAC Name: 1-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 530.58
Molecular Formula: C30H30N2O7
InChi Key: BYGKUWPLEGHFKX-ZRRKCSAHSA-N
SMILES: COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results