Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine, 98%

Catalog No. AAJ6596003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6596003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6596003 Supplier Thermo Scientific Chemicals Supplier No. J6596003
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23669-79-6 | C30H30N2O7 | 530.58 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in acetonitrile at 20mg/ml

Notes
Store in freezer (-20°)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23669-79-6
Molecular Formula C30H30N2O7
Molecular Weight (g/mol) 530.58
MDL Number MFCD00063410
InChI Key BYGKUWPLEGHFKX-ZRRKCSAHSA-N
Synonym 5'-o-4,4'-dimethoxytrityl-2'-deoxyuridine, dmt-du, 2'-deoxy-5'-o-dmt-uridine, 2'-deoxy-5'-o-4,4'-dimethoxytrityl uridine, 1-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxyoxolan-2-yl-3h-pyrimidine-2,4-dione, 5'-o-bis 4-methoxyphenyl phenyl methyl-2'-deoxyuridine, uridine,5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy, uridine, 5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy, 1-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenylmethoxy methyl-4-hydroxyoxolan-2-yl pyrimidine-2,4-dione
PubChem CID 90230
IUPAC Name 1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]pyrimidine-2,4-dione
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in acetonitrile at 20mg/ml
Formula Weight 530.57
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 245-814-9
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.