accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23677-93-2

IUPAC Name: copper(2+) bis(1,1,1-trifluoro-4-oxopent-2-en-2-olate)
Synonyms: copper(2+) bis(1,1,1-trifluoro-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 369.71
Molecular Formula: C10H8CuF6O4
InChi Key: GZVJAFMHAGQIEB-UHFFFAOYSA-L
SMILES: [Cu++].CC(=O)C=C([O-])C(F)(F)F.CC(=O)C=C([O-])C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results