Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trifluoro-2,4-pentanedionato)copper(II) 98.0+%, TCI America™
SDP

Catalog No. T07525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07525G Supplier TCI America Supplier No. T07525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23677-93-2
Molecular Formula C10H8CuF6O4
Molecular Weight (g/mol) 369.706
MDL Number MFCD00042511
InChI Key GZVJAFMHAGQIEB-GZBCLGFCSA-L
Synonym Copper(II) Trifluoroacetylacetonate, Trifluoroacetylacetono Copper(II) Salt
PubChem CID 101644372
IUPAC Name copper;(Z)-1,1,1-trifluoro-4-oxopent-2-en-2-olate;(E)-1,1,1-trifluoro-4-oxopent-2-en-2-olate
SMILES CC(=O)C=C(C(F)(F)F)[O-].CC(=O)C=C(C(F)(F)F)[O-].[Cu+2]

Specifications

Melting Point 195°C
Color Blue-Purple
Quantity 5 g
Formula Weight 369.71
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(trifluoro-2,4-pentanedionato)copper(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.