CAS RN 2368-53-8
IUPAC Name:
2,4-dichloro-1,3,5-trifluorobenzene
Synonyms:
2,4-dichloro-1,3,5-trifluorobenzene
Molecular Weight (g/mol):
200.97
Molecular Formula:
C6HCl2F3
InChi Key:
UOGVNYNJSZHQCN-UHFFFAOYSA-N
SMILES:
FC1=CC(F)=C(Cl)C(F)=C1Cl
1
–
1
of
1
results
1
1,3-Dichloro-2,4,6-trifluorobenzene 97.0+%, TCI America™
CAS: 2368-53-8 Molecular Formula: C6HCl2F3 Molecular Weight (g/mol): 200.97 MDL Number: MFCD00012242 InChI Key: UOGVNYNJSZHQCN-UHFFFAOYSA-N PubChem CID: 137563 IUPAC Name: 2,4-dichloro-1,3,5-trifluorobenzene SMILES: FC1=CC(F)=C(Cl)C(F)=C1Cl