Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-2,4,6-trifluorobenzene 97.0+%, TCI America™
SDP

Catalog No. D28685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28685G Supplier TCI America Supplier No. D28685G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2368-53-8
Molecular Formula C6HCl2F3
Molecular Weight (g/mol) 200.97
MDL Number MFCD00012242
InChI Key UOGVNYNJSZHQCN-UHFFFAOYSA-N
PubChem CID 137563
IUPAC Name 2,4-dichloro-1,3,5-trifluorobenzene
SMILES FC1=CC(F)=C(Cl)C(F)=C1Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 200.97
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dichloro-2,4,6-trifluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.