accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2370-63-0

H3COOOCH3CH2

CAS RN 2370-63-0

IUPAC Name: 2-ethoxyethyl 2-methylprop-2-enoate
Synonyms: ethoxyethyl methacrylate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: SFPNZPQIIAJXGL-UHFFFAOYSA-N
SMILES: CCOCCOC(=O)C(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results