Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. M1251500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1251500ML 500 mL
M125125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1251500ML Supplier TCI America Supplier No. M1251500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2370-63-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00009249
InChI Key SFPNZPQIIAJXGL-UHFFFAOYSA-N
Synonym 2-ethoxyethyl methacrylate, ethoxyethyl methacrylate, 2-propenoic acid, 2-methyl-, 2-ethoxyethyl ester, 2-ethoxy ethyl methacrylate, methacrylic acid, 2-ethoxyethyl ester, unii-5q6xr4mr8e, methacrylic acid 2-ethoxyethyl ester, 5q6xr4mr8e, 2-ethoxyethyl 2-methylacrylate, 2-ethyloxy ethyl 2-methylprop-2-enoate
PubChem CID 71404
IUPAC Name 2-ethoxyethyl 2-methylprop-2-enoate
SMILES CCOCCOC(=O)C(C)=C

Specifications

Color Colorless
Boiling Point 93°C
Quantity 500 mL
Formula Weight 158.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1049
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.