CAS RN 23702-49-0
CAS RN 23702-49-0
IUPAC Name:
2-oxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(14),4,6,8,10,12,15-heptaen-3-one
Synonyms:
2-oxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(14),4,6,8,10,12,15-heptaen-3-one
Molecular Weight (g/mol):
220.23
Molecular Formula:
C15H8O2
InChi Key:
SJXDHZGVKPNCHK-UHFFFAOYSA-N
SMILES:
O=C1OC2=CC=CC3=CC=C4C=CC=C1C4=C23
1
–
1
of
1
results
1
MP Biomedicals, Inc Benzo-1-naphthalide, MP Biomedicals
CAS: 23702-49-0 Molecular Formula: C15H8O2 Molecular Weight (g/mol): 220.23 InChI Key: SJXDHZGVKPNCHK-UHFFFAOYSA-N Synonym: 4-oxapyrene-5-one,5h-phenanthro 4,5-bcd pyran-5-one,ccris 2211,5-phenanthro 4,5-bcd pyran-5-one,benzo-1-naphthalide,4-oxa-pyrene-5-one,sjxdhzgvkpnchk-uhfffaoysa-n PubChem CID: 159957