Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo-1-naphthalide, MP Biomedicals

Catalog No. ICN209537 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209537 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209537 Supplier MP Biomedicals, Inc Supplier No. 0520953710

Chemical Identifiers

CAS 23702-49-0
Molecular Formula C15H8O2
Molecular Weight (g/mol) 220.23
InChI Key SJXDHZGVKPNCHK-UHFFFAOYSA-N
Synonym 4-oxapyrene-5-one, 5h-phenanthro 4,5-bcd pyran-5-one, ccris 2211, 5-phenanthro 4,5-bcd pyran-5-one, benzo-1-naphthalide, 4-oxa-pyrene-5-one, sjxdhzgvkpnchk-uhfffaoysa-n
PubChem CID 159957

Specifications

Quantity 10 g
Chemical Name or Material Benzo-1-naphthalide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.