accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23743-26-2

IUPAC Name: dicyclohexyl[2-(dicyclohexylphosphanyl)ethyl]phosphane
Synonyms: bis(dicyclohexylphosphino)ethane
Molecular Weight (g/mol): 422.62
Molecular Formula: C26H48P2
InChi Key: BOUYBUIVMHNXQB-UHFFFAOYSA-N
SMILES: C(CP(C1CCCCC1)C1CCCCC1)P(C1CCCCC1)C1CCCCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)