Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(dicyclohexylphosphino)ethane, 98%

Catalog No. AA3926904
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3926904 2 g
AA3926902 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3926904 Supplier Thermo Scientific Chemicals Supplier No. 03926904
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23743-26-2 | C26H48P2 | 422.618 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23743-26-2
Molecular Formula C26H48P2
Molecular Weight (g/mol) 422.618
MDL Number MFCD00015521
InChI Key BOUYBUIVMHNXQB-UHFFFAOYSA-N
Synonym 1,2-bis dicyclohexylphosphino ethane, ethylenebis dicyclohexylphosphine, phosphine, 1,2-ethanediylbis dicyclohexyl, 1,2-ethanediylbis dicyclohexyl phosphine, dicyclohexyl 2-dicyclohexylphosphanylethyl phosphane, bis 1,2-dicyclohexylphosphino ethane, pubchem6558, acmc-1capw, ethane-1,2-diylbis dicyclohexylphosphane, phosphine, 1,2-ethanediylbis*dicyclohexyl
PubChem CID 534202
IUPAC Name dicyclohexyl(2-dicyclohexylphosphanylethyl)phosphane
SMILES C1CCC(CC1)P(CCP(C2CCCCC2)C3CCCCC3)C4CCCCC4

Specifications

Quantity 2 g
UN Number UN1325
Sensitivity Air Sensitive
Formula Weight 422.61
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1,2-Bis(dicyclohexylphosphino)ethane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.