accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNNH2

CAS RN 23764-31-0

IUPAC Name: (3-aminopropyl)(propyl)amine
Synonyms: (3-aminopropyl)(propyl)amine
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: OWKYZAGJTTTXOK-UHFFFAOYSA-N
SMILES: CCCNCCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results