accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 23786-13-2

IUPAC Name: methyl 2-(4-methylphenyl)acetate
Synonyms: methyl 2-(4-methylphenyl)acetate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: LSAGWGNECLEVPE-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results