Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl p-Tolylacetate 99.0+%, TCI America™
SDP

Catalog No. M078325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M078325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M078325G Supplier TCI America Supplier No. M078325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23786-13-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00056243
InChI Key LSAGWGNECLEVPE-UHFFFAOYSA-N
Synonym methyl p-tolylacetate, methyl 4-methylphenylacetate, methyl 2-4-methylphenyl acetate, methyl 2-p-tolyl acetate, methylp-tolylacetate, p-methylphenylacetic acid methyl ester, methyl 4-methylphenyl acetate, p-tolylacetic acid methyl ester, 4-methylphenylacetic acid methyl ester, benzeneacetic acid, 4-methyl-, methyl ester
PubChem CID 90265
IUPAC Name methyl 2-(4-methylphenyl)acetate
SMILES COC(=O)CC1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 113°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl p-Tolylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.