accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2379-56-8

IUPAC Name: 6-nitro-1,2,3,4-tetrahydroquinoxaline-2,3-dione
Synonyms: 6-nitro-1,4-dihydroquinoxaline-2,3-dione
Molecular Weight (g/mol): 207.15
Molecular Formula: C8H5N3O4
InChi Key: RYMLSFWVYNAKAR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NC(=O)C(=O)NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results