Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dihydro-6-nitroquinoxaline-2,3-dione 95.0+%, TCI America™
SDP

Catalog No. D32415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32415G Supplier TCI America Supplier No. D32415G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2379-56-8
Molecular Formula C8H5N3O4
Molecular Weight (g/mol) 207.145
InChI Key RYMLSFWVYNAKAR-UHFFFAOYSA-N
Synonym 6-Nitro-1,4-dihydroquinoxaline-2,3-dione, 6-Nitroquinoxaline-2,3-dione
PubChem CID 4038142
IUPAC Name 6-nitro-1,4-dihydroquinoxaline-2,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])NC(=O)C(=O)N2

Specifications

Color Yellow
Quantity 5 g
Formula Weight 207.15
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dihydro-6-nitroquinoxaline-2,3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.