accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2381-45-5

H3COO(R)(S)(R)(S)OH(R)H3C(S)(S)H3CH3CO

CAS RN 2381-45-5

IUPAC Name: (1R,3aS,3bR,7S,9aR,9bS,11aS)-1-acetyl-7-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Synonyms: (1R,3aS,3bR,7S,9aR,9bS,11aS)-1-acetyl-7-hydroxy-9a,11a-dimethyl-2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9bH,10H,11H-cyclopenta[a]phenanthren-1-yl acetate
Molecular Weight (g/mol): 374.52
Molecular Formula: C23H34O4
InChi Key: KRBMZSYRIJAAMS-KIYFCUGLSA-N
SMILES: CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results