Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

17α-Acetoxypregnenolone, Thermo Scientific™

Catalog No. AC200990010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC200990010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC200990010 Supplier Thermo Scientific Chemicals Supplier No. 200990010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2381-45-5
Molecular Formula C23H34O4
Molecular Weight (g/mol) 374.521
InChI Key KRBMZSYRIJAAMS-KIYFCUGLSA-N
Synonym 17alpha-acetoxy pregnenolone, 3-hydroxy-20-oxo-5-pregnen-17alpha-yl acetate, 17a-acetoxy pregnenolone, 17alpha-acetoxypregnenolone, 17,a-acetoxy pregnenolone, 17-alpha-hydroxypregnenolone 17-acetate, 17-acetoxy-3, a-hydroxypregn-5-en-20-one, 17alpha-acetoxy-3beta-hydroxypregn-5-en-20-one, 3,a,17-dihydroxypregn-5-en-20-one 17-acetate, 3,a-17-acetyloxy-3-hydroxypregn-5-en-20-one
PubChem CID 102250
IUPAC Name [(3S,8R,9S,10R,13S,14S,17R)-17-acetyl-3-hydroxy-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)C1(CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)OC(=O)C

Specifications

Quantity 1 g
Formula Weight 374.51
Chemical Name or Material 17α-Acetoxypregnenolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.