accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23868-34-0

H3CNCH3H3CH3CHFF

CAS RN 23868-34-0

IUPAC Name: tetrabutylazanium hydrofluoride fluoride
Synonyms: tetrabutylammonium ion hydrofluoride fluoride
Molecular Weight (g/mol): 281.48
Molecular Formula: C16H37F2N
InChi Key: ZHBDKVWQJKYIFF-UHFFFAOYSA-M
SMILES: F.[F-].CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)