Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Spgs-550-M

Catalog No. 776033100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-603-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 77-603-3100ML Supplier Sigma Aldrich Supplier No. 776033100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Third Generation Designer Surfactant, in collaboration with the Lipshutz Group, is the most cost-effective surfactant to this point. Nok provides comparable yields for several cross-coupling reactions to first generation PTS (698717) and second generation TPGS (733857) at room temperature.Nok:A Phytosterol-Based Amphiphile Enabling Transition-Metal-Catalyzed Couplings in Water at Room Temperature

Specifications

CAS 23868-34-0
Molecular Formula C16H37F2N
Linear Formula (CH3CH2CH2CH2)4N(HF2)
Quantity 100 mL
MDL Number MFCD00077877
Synonym TBABF; Tetrabutylammonium bifluoride; Tetrabutylammonium difluoride
Molecular Weight (g/mol) 281.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.