CAS RN 2387505-50-0
IUPAC Name:
(1S,9S,10S)-17-(cyclopropylmethyl)-3,10-dimethoxy-17-azatetracyclo[7.5.3.0¹,¹⁰.0²,⁷]heptadeca-2(7),3,5-trien-13-one hydrochloride
Synonyms:
(1S,9S,10S)-17-(cyclopropylmethyl)-3,10-dimethoxy-17-azatetracyclo[7.5.3.0¹,¹⁰.0²,⁷]heptadeca-2(7),3,5-trien-13-one hydrochloride
Molecular Weight (g/mol):
391.94
Molecular Formula:
C22H30ClNO3
InChi Key:
MMOBYZBXXHEAAX-NCBCLDNOSA-N
SMILES:
Cl.[H][C@@]12CC3=C(C(OC)=CC=C3)[C@]3(CCN1CC1CC1)CC(=O)CC[C@@]23OC
1
–
1
of
1
results
1
R&D Systems™ Cyprodime hydrochloride
CAS: 2387505-50-0 Molecular Formula: C22H30ClNO3